2,4-Dichlor-5-sulfamoylbenzoesäure

Lieferant: FLUOROCHEM

Synonyme: Lasamide

014653-1G
FLUO014653-1GEA 0 EUR
FLUO014653-1G
2,4-Dichlor-5-sulfamoylbenzoesäure
2,4-Dichlor-5-sulfamoylbenzoesäure
Formel: C₇H₅Cl₂NO₄S
Molecular Weight: 270,09 g/mol
MDL: MFCD00007931
CAS-Nummer: 2736-23-4
EINECS: 220-358-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR