2,4-Dimethylbenzoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dimethylbenzoesäurechlorid

014685-1G
FLUO014685-1GEA 65.3 EUR
FLUO014685-1G
2,4-Dimethylbenzoylchlorid
2,4-Dimethylbenzoylchlorid
Formel: C₉H₉ClO
Molecular Weight: 168,62 g/mol
MDL: MFCD00798005
CAS-Nummer: 21900-42-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR