2,3-Dimethoxybenzoesäurechlorid

Lieferant: FLUOROCHEM

014690-1G
FLUO014690-1GEA 588 EUR
FLUO014690-1G
2,3-Dimethoxybenzoesäurechlorid
2,3-Dimethoxybenzoesäurechlorid
Formel: C₉H₉ClO₃
Molecular Weight: 200,62 g/mol
MDL: MFCD03424705
CAS-Nummer: 7169-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR