2,3,5-Triiodbenzoylchlorid

Lieferant: FLUOROCHEM

014701-1G
FLUO014701-1GEA 0 EUR
FLUO014701-1G
2,3,5-Triiodbenzoylchlorid
2,3,5-Triiodbenzoylchlorid
Formel: C₇H₂ClI₃O
Molecular Weight: 518,26 g/mol
MDL: MFCD01859916
CAS-Nummer: 42860-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR