3-Thienylacetylchlorid

Lieferant: FLUOROCHEM

014731-1G
FLUO014731-1GEA 0 EUR
FLUO014731-1G
3-Thienylacetylchlorid
3-Thienylacetylchlorid
Formel: C₆H₅ClOS
CAS-Nummer: 13781-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR