1-(3-Chlorbenzyl)piperazin
Lieferant: FLUOROCHEM
014884-1G
FLUO014884-1GEA
0
EUR
FLUO014884-1G
1-(3-Chlorbenzyl)piperazin
1-(3-Chlorbenzyl)piperazin
Formel:
C₁₁H₁₅ClN₂ Molecular Weight: 210,71 g/mol |
MDL:
MFCD02048053 CAS-Nummer: 23145-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...