1-(2,4-Dichlorbenzyl)piperazin
Lieferant: FLUOROCHEM
014885-1G
FLUO014885-1GEA
0
EUR
FLUO014885-1G
1-(2,4-Dichlorbenzyl)piperazin
1-(2,4-Dichlorbenzyl)piperazin
Formel:
C₁₁H₁₄Cl₂N₂ Molecular Weight: 245,15 g/mol |
MDL:
MFCD03407487 CAS-Nummer: 51619-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...