1-(3,4-Dichlorbenzyl)piperazin
Lieferant: FLUOROCHEM
014886-1G
FLUO014886-1GEA
0
EUR
FLUO014886-1G
1-(3,4-Dichlorbenzyl)piperazin
1-(3,4-Dichlorbenzyl)piperazin
Formel:
C₁₁H₁₄Cl₂N₂ Molecular Weight: 245,15 g/mol |
MDL:
MFCD03407488 CAS-Nummer: 55513-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...