1-(4-Brombenzyl)piperazin

Lieferant: FLUOROCHEM

014888-1G
FLUO014888-1GEA 0 EUR
FLUO014888-1G
1-(4-Brombenzyl)piperazin
1-(4-Brombenzyl)piperazin
Formel: C₁₁H₁₅BrN₂
Molecular Weight: 255,16 g/mol
MDL: MFCD02177416
CAS-Nummer: 91345-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR