4-Phenoxybenzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
4-Phenoxybenzolsulfochlorid, 4-Phenoxybenzol-1-sulfonylchlorid, 4-Phenoxybenzolsulfonsäurechlorid
014906-1G
FLUO014906-1GEA
130
EUR
FLUO014906-1G
4-Phenoxybenzolsulfonylchlorid
4-Phenoxybenzolsulfonylchlorid
Formel:
C₁₂H₉ClO₃S Molecular Weight: 268,72 g/mol Siedepunkt: 180…190 °C (6 mmHg) Schmelzpunkt: 44 °C |
MDL:
MFCD00625748 CAS-Nummer: 1623-92-3 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...