2-Amino-N-propylbenzamid
Lieferant: FLUOROCHEM
015202-1G
FLUO015202-1GEA
152
EUR
FLUO015202-1G
2-Amino-N-propylbenzamid
2-Amino-N-propylbenzamid
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178,231 g/mol |
MDL:
MFCD02374350 CAS-Nummer: 56814-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...