2-Amino-N-butylbenzamid

Lieferant: FLUOROCHEM

015203-1G
FLUO015203-1GEA 152 EUR
FLUO015203-1G
2-Amino-N-butylbenzamid
2-Amino-N-butylbenzamid
Formel: C₁₁H₁₆N₂O
Molecular Weight: 192,2600 g/mol
MDL: MFCD00456225
CAS-Nummer: 10494-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR