3-Iod-4-methoxybenzaldehyd

Lieferant: FLUOROCHEM

015239-1G
FLUO015239-1GEA 97.2 EUR
FLUO015239-1G
3-Iod-4-methoxybenzaldehyd
3-Iodo-4-methoxybenzaldehyde
Formel: C₈H₇IO₂
Molecular Weight: 262,04 g/mol
MDL: MFCD01009500
CAS-Nummer: 2314-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR