(2-Formylphenoxy)acetamid
Lieferant: FLUOROCHEM
015240-1G
FLUO015240-1GEA
471
EUR
FLUO015240-1G
(2-Formylphenoxy)acetamid
(2-Formylphenoxy)acetamid
Formel:
C₉H₉NO₃ |
MDL:
MFCD02257374 CAS-Nummer: 24590-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...