Imidazo[1,2-a]pyridin-2-carbonsäure Hydrobromid

Lieferant: FLUOROCHEM

015246-1G
FLUO015246-1GEA 378 EUR
FLUO015246-1G
Imidazo[1,2-a]pyridin-2-carbonsäure Hydrobromid
Imidazo[1,2-a]pyridin-2-carbonsäure Hydrobromid
Formel: C₈H₇BrN₂O₂
MDL: MFCD03425066
CAS-Nummer: 820245-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR