2-(2-Chloracetyl)thiophen

Lieferant: FLUOROCHEM

015347-1G
FLUO015347-1GEA 0 EUR
FLUO015347-1G
2-(2-Chloracetyl)thiophen
2-(2-Chloracetyl)thiophen
Formel: C₆H₅ClOS
CAS-Nummer: 29683-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR