1-(3-Bromphenyl)pyrrol

Lieferant: FLUOROCHEM

Synonyme: N-(3-Bromphenyl)pyrrol

015378-1G
FLUO015378-1GEA 228 EUR
FLUO015378-1G
1-(3-Bromphenyl)pyrrol
1-(3-Bromophenyl)pyrrole
Formel: C₁₀H₈BrN
Molecular Weight: 222,08 g/mol
MDL: MFCD02665243
CAS-Nummer: 107302-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR