8-Amino-5-bromchinolin

Lieferant: FLUOROCHEM

Synonyme: 5-Bromoquinolin-8-amine

015547-1G
FLUO015547-1GEA 136 EUR
FLUO015547-1G
8-Amino-5-bromchinolin
8-Amino-5-bromoquinoline
Formel: C₉H₇BrN₂
Molecular Weight: 223,0693 g/mol
MDL: MFCD01571832
CAS-Nummer: 53472-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR