4-Brom-3-sulfamoylbenzoesäure

Lieferant: FLUOROCHEM

015564-1G
FLUO015564-1GEA 747 EUR
FLUO015564-1G
4-Brom-3-sulfamoylbenzoesäure
4-Brom-3-sulfamoylbenzoesäure
Formel: C₇H₆BrNO₄S
CAS-Nummer: 59815-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR