4-Methylsalicylaldehyd
Lieferant: FLUOROCHEM
Synonyme:
2-Hydroxy-4-methylbenzaldehyde
015626-1G
FLUO015626-1GEA
42.9
EUR
FLUO015626-1G
4-Methylsalicylaldehyd
4-Methylsalicylaldehyd
Formel:
C₈H₈O₂ Molecular Weight: 136,15 g/mol |
MDL:
MFCD00799550 CAS-Nummer: 698-27-1 EINECS: 211-813-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...