2-Chlor-4'-methoxyacetanilid

Lieferant: FLUOROCHEM

Synonyme: 2-Chloro-N-(4-methoxyphenyl)acetamide

015705-1G
FLUO015705-1GEA 76.2 EUR
FLUO015705-1G
2-Chlor-4'-methoxyacetanilid
2-Chlor-4'-methoxyacetanilid
Formel: C₉H₁₀ClNO₂
MDL: MFCD00018907
CAS-Nummer: 22303-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR