2,7-Dimethylimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: FLUOROCHEM

CFF1057.1 CFF1057.5 CFF1057.25 015748-1G
CHMPCFF1057.1EA 522 EUR
CHMPCFF1057.1 CHMPCFF1057.5 CHMPCFF1057.25 FLUO015748-1G
2,7-Dimethylimidazo[1,2-a]pyridin-3-carbaldehyd
2,7-Dimethylimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₁₀H₁₀N₂O
MDL: MFCD03788999
CAS-Nummer: 820245-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR