2,8-Dimethylimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: FLUOROCHEM

CFF1058.1 CFF1058.5 015749-1G
CHMPCFF1058.1EA 522 EUR
CHMPCFF1058.1 CHMPCFF1058.5 FLUO015749-1G
2,8-Dimethylimidazo[1,2-a]pyridin-3-carbaldehyd
2,8-Dimethylimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₁₀H₁₀N₂O
MDL: MFCD03789000
CAS-Nummer: 820845-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR