4,5-Pyridazindicarbonsäure

Lieferant: FLUOROCHEM

016059-1G
FLUO016059-1GEA 143 EUR
FLUO016059-1G
4,5-Pyridazindicarbonsäure
Pyridazine-4,5-dicarboxylic acid
Formel: C₆H₄N₂O₄
MDL: MFCD06742784
CAS-Nummer: 59648-14-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR