2-(1H-Pyrazol-4-yl)pyridin

Lieferant: FLUOROCHEM

016083-1G 016083-5G
FLUO016083-1GEA 107 EUR
FLUO016083-1G FLUO016083-5G
2-(1H-Pyrazol-4-yl)pyridin
2-(1H-Pyrazol-4-yl)pyridin
Formel: C₈H₇N₃
Molecular Weight: 145.1613 g/mol
MDL: MFCD02579318
CAS-Nummer: 439106-75-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR