Ethyl-2-chlor-4-oxazolcarboxylat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-chlorooxazole-4-carboxylate

016119-1G
FLUO016119-1GEA 56.1 EUR
FLUO016119-1G
Ethyl-2-chlor-4-oxazolcarboxylat
Ethyl-2-chlor-4-oxazolcarboxylat
Formel: C₆H₆ClNO₃
Molecular Weight: 175,57 g/mol
MDL: MFCD06660120
CAS-Nummer: 460081-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR