(1H-Imidazol-4-yl)methanol Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: 4-Imidazolemethanol hydrochloride, 4(5)-Imidazolemethanol hydrochloride, 4-(Hydroxymethyl)imidazole hydrochloride

016121-1G 016121-5G
FLUO016121-1GEA 42.6 EUR
FLUO016121-1G FLUO016121-5G
(1H-Imidazol-4-yl)methanol Hydrochlorid
(1H-Imidazol-4-yl)methanol Hydrochlorid
Formel: C₄H₇ClN₂O
MDL: MFCD00012697
CAS-Nummer: 32673-41-9
EINECS: 251-150-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR