Ethyl-2-amino-4-oxazolcarboxylat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-aminooxazole-4-carboxylate

016128-1G
FLUO016128-1GEA 30.8 EUR
FLUO016128-1G
Ethyl-2-amino-4-oxazolcarboxylat
Ethyl-2-amino-4-oxazolcarboxylat
Formel: C₆H₈N₂O₃
Molecular Weight: 156.14 g/mol
MDL: MFCD06659907
CAS-Nummer: 177760-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR