2-Brom-4-methylthiazol

Lieferant: FLUOROCHEM

016134-1G 016134-5G
FLUO016134-1GEA 57.8 EUR
FLUO016134-1G FLUO016134-5G
2-Brom-4-methylthiazol
2-Bromo-4-methylthiazole
Formel: C₄H₄BrNS
Molecular Weight: 178.05 g/mol
MDL: MFCD06660130
CAS-Nummer: 7238-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR