2-Brom-4-methylthiazol
Lieferant: FLUOROCHEM
016134-1G
016134-5G
FLUO016134-1GEA
57.8
EUR
FLUO016134-1G
FLUO016134-5G
2-Brom-4-methylthiazol
2-Bromo-4-methylthiazole
Formel:
C₄H₄BrNS Molecular Weight: 178.05 g/mol |
MDL:
MFCD06660130 CAS-Nummer: 7238-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...