4-Benzyloxyindol
Lieferant: FLUOROCHEM
016140-1G
016140-5G
FLUO016140-1GEA
31.1
EUR
FLUO016140-1G
FLUO016140-5G
4-Benzyloxyindol
4-Benzyloxyindol
Formel:
C₁₅H₁₃NO |
MDL:
MFCD00047200 CAS-Nummer: 20289-26-3 EINECS: 243-690-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...