3-Carboxy-5-nitrophenylboronsäure

Lieferant: FLUOROCHEM

016141-1G 016141-5G
FLUO016141-1GEA 31.1 EUR
FLUO016141-1G FLUO016141-5G
3-Carboxy-5-nitrophenylboronsäure
3-Carboxy-5-nitrophenylboronsäure
Formel: C₇H₆BNO₆
MDL: MFCD00757433
CAS-Nummer: 101084-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR