3-Brom-5-(chlormethyl)isoxazol

Lieferant: FLUOROCHEM

016144-1G
FLUO016144-1GEA 599 EUR
FLUO016144-1G
3-Brom-5-(chlormethyl)isoxazol
3-Bromo-5-(chloromethyl)isoxazole
Formel: C₄H₃BrClNO
MDL: MFCD06660137
CAS-Nummer: 124498-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR