2-Brom-5-formylthiazol

Lieferant: FLUOROCHEM

Synonyme: 2-Bromothiazole-5-carboxaldehyde

016151-1G
FLUO016151-1GEA 79.1 EUR
FLUO016151-1G
2-Brom-5-formylthiazol
2-Brom-5-formylthiazol
Formel: C₄H₂BrNOS
Molecular Weight: 192,03 g/mol
MDL: MFCD03788567
CAS-Nummer: 464192-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR