2-Brom-5-thiazolcarbonsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Bromothiazole-5-carboxylic acid
016153-5G
FLUO016153-5GEA
94.1
EUR
FLUO016153-5G
2-Brom-5-thiazolcarbonsäure
2-Bromo-5-thiazolecarboxylic acid
Formel:
C₄H₂BrNO₂S Molecular Weight: 208,04 g/mol |
MDL:
MFCD04115730 CAS-Nummer: 54045-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...