7-Indolcarbaldehyd
Lieferant: FLUOROCHEM
Synonyme:
Indol-7-carboxaldehyd
016158-1G
FLUO016158-1GEA
48.8
EUR
FLUO016158-1G
7-Indolcarbaldehyd
7-Indolcarbaldehyd
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol |
MDL:
MFCD01318152 CAS-Nummer: 1074-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...