4-(Hydroxymethyl)indol
Lieferant: FLUOROCHEM
Synonyme:
(1H-Indol-4-yl)methanol
016160-1G
FLUO016160-1GEA
195
EUR
FLUO016160-1G
4-(Hydroxymethyl)indol
4-(Hydroxymethyl)indole
Formel:
C₉H₉NO Molecular Weight: 147,1739 g/mol |
MDL:
MFCD01632220 CAS-Nummer: 1074-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...