5-Thiazolcarbonsäure

Lieferant: FLUOROCHEM

016173-1G 016173-5G
FLUO016173-1GEA 45.8 EUR
FLUO016173-1G FLUO016173-5G
5-Thiazolcarbonsäure
5-Thiazolecarboxylic acid
Formel: C₄H₃NO₂S
Molecular Weight: 129.1371 g/mol
MDL: MFCD03428539
CAS-Nummer: 14527-41-4
EINECS: 238-545-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR