5-Thiazolcarbonsäure
Lieferant: FLUOROCHEM
016173-1G
016173-5G
FLUO016173-1GEA
45.8
EUR
FLUO016173-1G
FLUO016173-5G
5-Thiazolcarbonsäure
5-Thiazolecarboxylic acid
Formel:
C₄H₃NO₂S Molecular Weight: 129.1371 g/mol |
MDL:
MFCD03428539 CAS-Nummer: 14527-41-4 EINECS: 238-545-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...