6-(Trifluormethyl)oxindol

Lieferant: FLUOROCHEM

CFF1287. 016176-1G
CHMPCFF1287.EA 0 EUR
CHMPCFF1287. FLUO016176-1G
6-(Trifluormethyl)oxindol
6-(Trifluormethyl)oxindol
Formel: C₉H₆F₃NO
MDL: MFCD01075220
CAS-Nummer: 1735-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR