3-Brom-5-(hydroxymethyl)isoxazol
Lieferant: FLUOROCHEM
Synonyme:
(3-Bromoisoxazol-5-yl)methanol
016180-1G
FLUO016180-1GEA
219
EUR
FLUO016180-1G
3-Brom-5-(hydroxymethyl)isoxazol
3-Bromo-5-(hydroxymethyl)isoxazole
Formel:
C₄H₄BrNO₂ Molecular Weight: 177,98 g/mol |
MDL:
MFCD04035586 CAS-Nummer: 25742-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...