3-Brom-5-(4-fluorphenyl)isoxazol

Lieferant: FLUOROCHEM

016186-500MG
FLUO016186-500MGEA 447 EUR
FLUO016186-500MG
3-Brom-5-(4-fluorphenyl)isoxazol
3-Bromo-5-(4-fluorophenyl)isoxazole
Formel: C₉H₅BrFNO
MDL: MFCD06660142
CAS-Nummer: 903130-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR