5-Brom-2-fluorpyrimidin
Lieferant: FLUOROCHEM
016201-1G
FLUO016201-1GEA
45.9
EUR
FLUO016201-1G
5-Brom-2-fluorpyrimidin
5-Bromo-2-fluoropyrimidine
Formel:
C₄H₂BrFN₂ |
MDL:
MFCD07787364 CAS-Nummer: 62802-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...