2-Chlor-5-formylphenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
(2-Chloro-5-formylphenyl)boronic acid
016204-1G
FLUO016204-1GEA
52.7
EUR
FLUO016204-1G
2-Chlor-5-formylphenylboronsäure
2-Chlor-5-formylphenylboronsäure
Formel:
C₇H₆BClO₃ Molecular Weight: 184,3847 g/mol |
MDL:
MFCD06656269 CAS-Nummer: 1150114-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...