2-Fluor-5-nitrophenylboronsäure

Lieferant: FLUOROCHEM

016221-1G 016221-5G
FLUO016221-1GEA 39.6 EUR
FLUO016221-1G FLUO016221-5G
2-Fluor-5-nitrophenylboronsäure
2-Fluor-5-nitrophenylboronsäure
Formel: C₆H₅BFNO₄
MDL: MFCD03092929
CAS-Nummer: 819849-20-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR