5-Hydroxyindol

Lieferant: FLUOROCHEM

Synonyme: 1H-Indol-5-ol, 5-Indolol, Indol-5-ol, 5-Hydroxy-1H-indol

016237-1G
FLUO016237-1GEA 45.9 EUR
FLUO016237-1G
5-Hydroxyindol
5-Hydroxyindol
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 107…108 °C
MDL: MFCD00005677
CAS-Nummer: 1953-54-4
EINECS: 217-782-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR