1-Boc-5-fluor-2-indolboronsäure

Lieferant: FLUOROCHEM

Synonyme: 1-Boc-5-fluoroindole-2-boronic Acid, N-(Boc)-5-fluoroindole-2-boronic acid

016247-1G
FLUO016247-1GEA 288 EUR
FLUO016247-1G
1-Boc-5-fluor-2-indolboronsäure
1-Boc-5-fluor-2-indolboronsäure
Formel: C₁₃H₁₅BFNO₄
Molecular Weight: 279,07 g/mol
MDL: MFCD04114581
CAS-Nummer: 352359-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR