3-(Hydroxymethyl)phenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(Hydroxymethyl)benzeneboronic acid, (3-Hydroxymethyl)phenylboronic acid
016258-1G
016258-25G
FLUO016258-1GEA
31.1
EUR
FLUO016258-1G
FLUO016258-25G
3-(Hydroxymethyl)phenylboronsäure
3-(Hydroxymethyl)phenylboronsäure
Formel:
C₇H₉BO₃ |
MDL:
MFCD01317846 CAS-Nummer: 87199-15-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...