3-(Hydroxymethyl)phenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(Hydroxymethyl)benzeneboronic acid, (3-Hydroxymethyl)phenylboronic acid

016258-1G 016258-25G
FLUO016258-1GEA 31.1 EUR
FLUO016258-1G FLUO016258-25G
3-(Hydroxymethyl)phenylboronsäure
3-(Hydroxymethyl)phenylboronsäure
Formel: C₇H₉BO₃
MDL: MFCD01317846
CAS-Nummer: 87199-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR