4-(Hydroxymethyl)phenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-(Hydroxymethyl)benzeneboronic acid

016259-1G 016259-25G 016259-5G
FLUO016259-1GEA 31.1 EUR
FLUO016259-1G FLUO016259-25G FLUO016259-5G
4-(Hydroxymethyl)phenylboronsäure
4-(Hydroxymethyl)phenylboronsäure
Formel: C₇H₉BO₃
MDL: MFCD00792672
CAS-Nummer: 59016-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR